Anti-FRS2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315113EA 564 EUR
ORIGTA315113
Anti-FRS2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: FRS2 (fibroblast growth factor receptor substrate 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR