Imprimer

Anti-ACOT2 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA315131EA 564 EUR
ORIGTA315131
Anti-ACOT2 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: ACOT2 (acyl-CoA thioesterase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR