Imprimer

Anti-LILRB4 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA315136EA 564 EUR
ORIGTA315136
Anti-LILRB4 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: LILRB4 (leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR