Anti-BVES Rabbit Polyclonal Antibody

Supplier: OriGene
TA315155
ORIGTA315155EA 564 EUR
ORIGTA315155
Anti-BVES Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: BVES (blood vessel epicardial substance)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR