Anti-IKZF2 Rabbit Polyclonal Antibody

Supplier: OriGene
TA315181
ORIGTA315181EA 564 EUR
ORIGTA315181
Anti-IKZF2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: IKZF2 (IKAROS family zinc finger 2 (Helios))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR