Anti-SLC4A1AP Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315208EA 532 EUR
ORIGTA315208
Anti-SLC4A1AP Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: SLC4A1AP (solute carrier family 4 (anion exchanger), member 1, adaptor protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR