Anti-ZC3H13 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315219EA 543 EUR
ORIGTA315219
Anti-ZC3H13 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: ZC3H13 (zinc finger CCCH-type containing 13)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR