Imprimer

Anti-MPRIP Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA315224EA 564 EUR
ORIGTA315224
Anti-MPRIP Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: MPRIP (myosin phosphatase Rho interacting protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR