Imprimer
Anti-CSTF2T Rabbit Polyclonal Antibody
Fournisseur: OriGene
ORIGTA315235EA 543 EUR
ORIGTA315235
Anti-CSTF2T Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000
Type: Primary
Antigen: CSTF2T (cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa, tau variant)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant