Imprimer

Anti-RRP8 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA315242EA 564 EUR
ORIGTA315242
Anti-RRP8 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: RRP8 (ribosomal RNA processing 8, methyltransferase, homolog (yeast))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR