Anti-MED13L Rabbit Polyclonal Antibody

Supplier: OriGene
TA315244
ORIGTA315244EA 564 EUR
ORIGTA315244
Anti-MED13L Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: MED13L (mediator complex subunit 13-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR