Anti-EID1 Rabbit Polyclonal Antibody

Supplier: OriGene
TA315270
ORIGTA315270EA 564 EUR
ORIGTA315270
Anti-EID1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: EID1 (EP300 interacting inhibitor of differentiation 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR