Anti-SHC2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315284EA 564 EUR
ORIGTA315284
Anti-SHC2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: SHC2 (SHC (Src homology 2 domain containing) transforming protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR