Anti-RAB3GAP2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315285EA 552 EUR
ORIGTA315285
Anti-RAB3GAP2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: RAB3GAP2 (RAB3 GTPase activating protein subunit 2 (non-catalytic))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR