Anti-TNFAIP8 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315288EA 564 EUR
ORIGTA315288
Anti-TNFAIP8 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: TNFAIP8 (tumor necrosis factor, alpha-induced protein 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR