Imprimer

Anti-GIMAP2 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA315312EA 564 EUR
ORIGTA315312
Anti-GIMAP2 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: GIMAP2 (GTPase, IMAP family member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR