Imprimer
Anti-SLC39A1 Rabbit Polyclonal Antibody
Fournisseur: OriGene
ORIGTA315370EA 564 EUR
ORIGTA315370
Anti-SLC39A1 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000
Type: Primary
Antigen: SLC39A1 (solute carrier family 39 (zinc transporter), member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human