Anti-CDKL3 Rabbit Polyclonal Antibody

Supplier: OriGene
TA315458
ORIGTA315458EA 543 EUR
ORIGTA315458
Anti-CDKL3 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: CDKL3 (cyclin-dependent kinase-like 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR