Anti-MBTPS2 Rabbit Polyclonal Antibody

Supplier: OriGene
TA315468
ORIGTA315468EA 564 EUR
ORIGTA315468
Anti-MBTPS2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: MBTPS2 (membrane-bound transcription factor peptidase, site 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR