Imprimer

Anti-CHRAC1 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA315515EA 564 EUR
ORIGTA315515
Anti-CHRAC1 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:1000

Type: Primary
Antigen: CHRAC1 (chromatin accessibility complex 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR