Anti-MOV10L1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315523EA 564 EUR
ORIGTA315523
Anti-MOV10L1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: MOV10L1 (Mov10l1, Moloney leukemia virus 10-like 1, homolog (mouse))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR