Anti-FAM35A Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315529EA 564 EUR
ORIGTA315529
Anti-FAM35A Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: FAM35A (family with sequence similarity 35, member A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR