Imprimer

Anti-ELOVL2 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA315545EA 543 EUR
ORIGTA315545
Anti-ELOVL2 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: ELOVL2 (ELOVL fatty acid elongase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR