Anti-ATG16L1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315564EA 552 EUR
ORIGTA315564
Anti-ATG16L1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5000

Type: Primary
Antigen: ATG16L1 (autophagy related 16-like 1 (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR