Imprimer

Anti-SLC39A9 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA315582EA 564 EUR
ORIGTA315582
Anti-SLC39A9 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: SLC39A9 (solute carrier family 39, member 9)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR