Imprimer

Anti-POLR3B Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA315598EA 564 EUR
ORIGTA315598
Anti-POLR3B Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: POLR3B (polymerase (RNA) III (DNA directed) polypeptide B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR