Anti-LMO3 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315611EA 552 EUR
ORIGTA315611
Anti-LMO3 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: LMO3 (LIM domain only 3 (rhombotin-like 2))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR