Anti-UBFD1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315615EA 543 EUR
ORIGTA315615
Anti-UBFD1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: UBFD1 (ubiquitin family domain containing 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR