Imprimer

Anti-KCNK15 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA315706EA 564 EUR
ORIGTA315706
Anti-KCNK15 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: KCNK15 (potassium channel, subfamily K, member 15)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR