Imprimer

Anti-ABHD4 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA315707EA 543 EUR
ORIGTA315707
Anti-ABHD4 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: ABHD4 (abhydrolase domain containing 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR