Anti-TP53AIP1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315712EA 543 EUR
ORIGTA315712
Anti-TP53AIP1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:1000

Type: Primary
Antigen: TP53AIP1 (tumor protein p53 regulated apoptosis inducing protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR