Anti-MMP27 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315714EA 543 EUR
ORIGTA315714
Anti-MMP27 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: MMP27 (matrix metallopeptidase 27)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR