Imprimer

Anti-OR51B2 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA315782EA 564 EUR
ORIGTA315782
Anti-OR51B2 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: OR51B2 (olfactory receptor, family 51, subfamily B, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR