Anti-BCL2L14 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315784EA 543 EUR
ORIGTA315784
Anti-BCL2L14 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5000

Type: Primary
Antigen: BCL2L14 (BCL2-like 14 (apoptosis facilitator))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR