Imprimer

Anti-EPS8L3 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA315788EA 543 EUR
ORIGTA315788
Anti-EPS8L3 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: EPS8L3 (EPS8-like 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR