Anti-HSPBAP1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315797EA 543 EUR
ORIGTA315797
Anti-HSPBAP1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5000

Type: Primary
Antigen: HSPBAP1 (HSPB (heat shock 27kDa) associated protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR