Anti-CABLES2 Rabbit Polyclonal Antibody

Supplier: OriGene
TA315863
ORIGTA315863EA 564 EUR
ORIGTA315863
Anti-CABLES2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: CABLES2 (Cdk5 and Abl enzyme substrate 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR