Anti-CEP78 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315897EA 552 EUR
ORIGTA315897
Anti-CEP78 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: CEP78 (centrosomal protein 78kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR