Imprimer

Anti-ACBD6 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA315909EA 543 EUR
ORIGTA315909
Anti-ACBD6 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5000

Type: Primary
Antigen: ACBD6 (acyl-CoA binding domain containing 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR