Anti-SFT2D3 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315938EA 543 EUR
ORIGTA315938
Anti-SFT2D3 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: SFT2D3 (SFT2 domain containing 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR