Anti-VPS26B Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA316005EA 532 EUR
ORIGTA316005
Anti-VPS26B Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: VPS26B (vacuolar protein sorting 26 homolog B (S. pombe))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR