Anti-CMTM1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA316009EA 552 EUR
ORIGTA316009
Anti-CMTM1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:60000

Type: Primary
Antigen: CMTM1 (CKLF-like MARVEL transmembrane domain containing 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR