Anti-MRPL54 Rabbit Polyclonal Antibody

Supplier: OriGene
TA316026
ORIGTA316026EA 564 EUR
ORIGTA316026
Anti-MRPL54 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: MRPL54 (mitochondrial ribosomal protein L54)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR