Imprimer

Anti-OR56A1 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA316051EA 543 EUR
ORIGTA316051
Anti-OR56A1 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: OR56A1 (olfactory receptor, family 56, subfamily A, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR