Anti-TPH2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA316055EA 564 EUR
ORIGTA316055
Anti-TPH2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: TPH2 (tryptophan hydroxylase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR