Imprimer

Anti-SPIC Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA316056EA 543 EUR
ORIGTA316056
Anti-SPIC Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: SPIC (Spi-C transcription factor (Spi-1/PU.1 related))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR