Imprimer

Anti-CDYL2 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA316072EA 543 EUR
ORIGTA316072
Anti-CDYL2 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: CDYL2 (chromodomain protein, Y-like 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR