Imprimer

Anti-MRPL10 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA316074EA 564 EUR
ORIGTA316074
Anti-MRPL10 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: MRPL10 (mitochondrial ribosomal protein L10)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR