Anti-OR13C8 Rabbit Polyclonal Antibody

Supplier: OriGene
TA316115
ORIGTA316115EA 564 EUR
ORIGTA316115
Anti-OR13C8 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: OR13C8 (olfactory receptor, family 13, subfamily C, member 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR