Anti-CPNE8 Rabbit Polyclonal Antibody

Supplier: OriGene
TA316141
ORIGTA316141EA 564 EUR
ORIGTA316141
Anti-CPNE8 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: CPNE8 (copine VIII)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR